PloS one
Expression of inhibin-alpha is regulated synergistically by Wilms' tumor gene 1 (Wt1) and steroidogenic factor-1 (Sf1) in sertoli cells.
PMID 23326390
Abstract
Wt1 encodes a zinc finger nuclear transcriptional factor, which is specifically expressed in testicular Sertoli cells and knockdown of Wt1 in Sertoli cells causes male mice subfertility. However, the underlying mechanism is still unclear. In this study, we found that expression of inhibin-α is significantly reduced in Wt1-deficient Sertoli cells. Luciferase assays using the inhibin-α promoter indicated that the inhibin-α promoter is transactivated by the Wt1 A, and B isoforms (-KTS), but not the C, and D isoforms (+KTS). Analysis of the Wt1 responsive element of the inhibin-α promoter region using site-directed mutagenesis showed that the nucleotides between -58 and -49 are essential for Wt1-dependent transactivation of the inhibin-α promoter. ChIP assays indicated that Wt1 directly interacts with the inhibin-α promoter. In addition, the inhibin-α promoter is activated synergistically by Wt1 and Sf1. Mutation of the ligand binding domain (LBD) of Sf1 (residues 235-238) completely abolished the synergistic action between Wt1 and Sf1, but did not affect the physical interaction between these two proteins, suggesting that other factor(s) may also be involved in the regulation of inhibin-α in Sertoli cells. Further studies demonstrated that β-catenin enhances the synergistic activation of Wt1 and Sf1 on the inhibin-α promoter. Given the fact that inhibin-α, a subunit of inhibin, is known to be involved in the regulation of spermatogenesis and testicular steroidogenesis, this study reveals a new regulatory mechanism of inhibin-α in Sertoli cells and also sheds light on the physiological functions of Wt1 in gonad development and spermatogenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I9149 Inhibin from porcine ovary, lyophilized powder