EMAIL THIS PAGE TO A FRIEND
MMW Fortschritte der Medizin
[Glycosuria with SGLT-2 inhibitors--a new treatment approach for type 2 diabetes mellitus].
PMID 23326927
Abstract
A new mechanism of action in the form of sodium-glucose co-transporter-(SGLT-)2 inhibitors will be available shortly for the treatment of type 2 diabetic patients. Overview. Fasting and postprandial blood glucose and HbA(1c) concentrations are indirectly reduced by the inhibition of glucose reabsorption and increased glycosuria. SGLT-2 inhibitors also have a positive impact on body weight and blood pressure of type 2 diabetics. In the available registration trials conducted to date, the SGLT-2 inhibitors appeared overall as a safe class of drugs. The clinical importance of an increased incidence of genital infections--in particular in special patientpopulations--requires further clarification. Long-term trials are currently underway to verify safety and in particular cardiovascular effects of this drug class.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G117
Glipizide, solid
C21H27N5O4S