EMAIL THIS PAGE TO A FRIEND
Journal of the Medical Association of Thailand = Chotmaihet thangphaet
Comparison of the effectiveness in bacterial decontamination between chlorhexidine gluconate and povidone-iodine solution in foot and ankle: a pilot study.
PMID 23326989
Abstract
The decontamination for foot and ankle surgery should be considered as a special preparation due to higher rate of bacterial contamination. The footwear and humidity is also the issue of interest especially in tropical country. The contamination before surgery should be reduced to avoid the infection. The effectiveness of antiseptics and special condition for the foot and ankle surgery should be elucidated for better medical care. The twenty volunteers were included in the present study. In group 1, the foot was scrubbed with 7.5% Povidone-lodine and painted with 10% Povidone-lodine solution. In group II, the foot was scrubbed with Chlorhexidine gluconate scrub and painted with 2% Chlorhexidine gluconate in 70% alcohol. At the beginning and end of the preparation, specimens were taking from all toes, nailfold, interdigital web spaces. These samples were sent for aerobic bacterial cultures. The results were interpreted as positive or negative cultivation and the number of bacterial colonies. All of the samples from 40 feet were collected; In Group I, positive culture was 5 samples (25%). In Group II, positive culture was 2 samples (10%) (p = 0.2). The Chlorhexidine gluconate and Povidone-lodine are effective in reduction the number of bacterial colonization. The steps of preparation before surgery also play an important role in eliminating the pathogenic bacteria. Both antiseptics were found no significant different in efficacy of pathogenic bacteria reduction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PVP1
Poly(vinylpyrrolidone)–Iodine complex