EMAIL THIS PAGE TO A FRIEND
Journal of cardiovascular pharmacology
Domperidone and sudden cardiac death: How much longer should we wait?
PMID 23328387
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D2955000
Domperidone, European Pharmacopoeia (EP) Reference Standard
C22H24ClN5O2