EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Chemistry. Re-engineering nature's catalysts.
PMID 23329039
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
295183
Cyclopropane, ≥99%
C3H6