EMAIL THIS PAGE TO A FRIEND
Harefuah
[Thiopurine-induced hyperammonaemic encephalopathy in a patient with Crohn's disease].
PMID 23330262
Abstract
Thiopurine drugs, azathioprine (Imuran) and 6-mercaptopurine (6-MP), are immunomodulators that have been shown to be effective at inducing and maintaining remission in inflammatory bowel disease. Although usually well-tolerated, the occurrence of side effects, typically myelotoxicity and hepatotoxicity, is a major drawback. The side effects can be classified as dose-dependent and independent. Both cholestatic hepatitis and endothelial injury, leading to vascular congestion and nodular regenerative hyperplasia, have been described during therapy with thiopurines, which can end up with portal hypertension. These injuries are potentially mediated by different metabolites. In this article we present a case of hyperammonaemic encephalopathy during therapy with 6-MP, possibly the first recorded in the literature, which probably resulted from the combination of thiopurine-induced liver injury with portal hypertension and the presence of spontaneous portosystemic venous shunts.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
852678
6-Mercaptopurine monohydrate, 98%
C5H4N4S · H2O