EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Structure-based design and synthesis of C-1- and C-4-modified analogs of zanamivir as neuraminidase inhibitors.
PMID 23330908
Abstract
In order to exploit the 430-cavity in the active sites of neuraminidases, 22 zanamivir analogs with C-1 and C-4 modification were synthesized, and their inhibitory activities against both group-1 (H5N1, H1N1) and group-2 neuraminidases (H3N2) were determined. Compound 9f exerts the most potency, with IC(50) value of 0.013, 0.001, and 0.09 μM against H3N2, H5N1, and H1N1, which is similar to that of zanamivir (H3N2 IC(50) = 0.0014 μM, H5N1 IC(50) = 0.012 μM, H1N1 IC(50) = 0.001 μM). Pharmacokinetic studies of compound 9f in rats showed a much longer plasma half-life (t(1/2)) than that of zanamivir following administration (po dose). Molecular modeling provided information about the binding model between the new inhibitors and neuraminidase, with the elongated groups at the C-1-position being projected toward the 430-loop region. This study may represent a novel starting point for the future development of improved antiflu agents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N7271 α(2→3) Neuraminidase from Streptococcus pneumoniae, buffered aqueous solution
N5521 α(2→3,6) Neuraminidase from Clostridium perfringens (C. welchii), recombinant, expressed in E. coli, buffered aqueous solution, ≥250 units/mg protein
N3786
α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, Proteomics Grade, suitable for MALDI-TOF MS
N8271 α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, recombinant, expressed in E. coli, buffered aqueous solution
N5254 Neuraminidase Agarose from Clostridium perfringens (C. welchii), Type VI-A, ammonium sulfate suspension
N2876
Neuraminidase from Clostridium perfringens (C. welchii), Suitable for manufacturing of diagnostic kits and reagents, Type V, lyophilized powder
N3001
Neuraminidase from Clostridium perfringens (C. welchii), Type VI, lyophilized powder, 6-10 units/mg protein (using 4MU-NANA), 2-5 units/mg protein (mucin)
N5631
Neuraminidase from Clostridium perfringens (C. welchii), Type VIII, lyophilized powder, 10-20 units/mg protein (using 4MU-NANA), 3.5-8.0 units/mg protein (mucin)
N2133
Neuraminidase from Clostridium perfringens (C. welchii), Type X, lyophilized powder, ≥50 units/mg protein (using 4MU-NANA)
N7885
Neuraminidase from Vibrio cholerae, Type III, buffered aqueous solution, 0.2 μm filtered, 1-5 units/mg protein (Lowry, using NAN-lactose)
N6514
Neuraminidase from Vibrio cholerae, Type II, buffered aqueous solution, 8-24 units/mg protein (Lowry, using NAN-lactose)
72197
Neuraminidase from Vibrio cholerae, ≥1.5 U/mL, specific activity ≥1.5 U/mg protein