EMAIL THIS PAGE TO A FRIEND
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology
Allergen bronchoprovocation and chitinases in allergic asthma.
PMID 23331556