EMAIL THIS PAGE TO A FRIEND
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.]
Arbor cutis: hyperkeratotic plaque secondary to the use of imiquimod for the treatment of squamous cell carcinoma in situ.
PMID 23331869