EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
A novel method for the preparation of a neurotoxic complex.
PMID 23333268
Abstract
Neurodegenerative diseases such as Alzheimer's disease (AD) and Parkinson's disease (PD) can be attributed to the specific degeneration of neuronal cells in the brain. However, the natures and action modes of toxic species remain largely unknown. Here, we present a simple and fast method for the preparation of neurotoxic complex with α-synuclein, which is implicated in PD.