EMAIL THIS PAGE TO A FRIEND
Experimental eye research
Ocular and systemic toxicity of intravitreal topotecan in rabbits for potential treatment of retinoblastoma.
PMID 23333535
Abstract
Treatment of intraocular retinoblastoma with vitreous seeding is a challenge. Different routes of chemotherapy administration have been explored in order to attaining pharmacological concentrations into the posterior chamber. Intravitreal drug injection is a promissing route for maximum bioavailability to the vitreous but it requires a well defined dose for achieving tumor control while limited toxicity to the retina. Topotecan proved to be a promising agent for retinoblastoma treatment due to its pharmacological activity and limited toxicity. High and prolonged concentrations were achieved in the rabbit vitreous after 5 μg of intravitreal topotecan. However, whether a lower dose could achieve potentially therapeutic levels remained to be determined. Thus, we here study the pharmacokinetics of topotecan after 0.5 μg and the toxicity profile of intravitreal topotecan in the rabbit eye as a potential treatment of retinoblastoma. A cohort of rabbits was used to study topotecan disposition in the vitreous after a single dose of 0.5 μg of intravitreal topotecan. In addition, an independent cohort of non-tumor bearing rabbits was employed to evaluate the clinical and retinal toxicity after four weekly injections of two different doses of intravitreal topotecan (Group A, 5 μg/dose; Group B, 0.5 μg/dose) to the right eye of each animal. The same volume (0.1 ml) of normal saline was administered to the left eye as control. A third group of rabbits (Group C) served as double control (both eyes injected with normal saline). Animals were weekly evaluated for clinical and hematologic values and ocular evaluations were performed with an inverse ophthalmoscope to establish potential topotecan toxicity. Weekly controls included topotecan quantitation in plasma of all rabbits. Electroretinograms (ERGs) were recorded before and after topotecan doses. One week after the last injection, topotecan concentrations were measured in vitreous of all eyes and samples for retinal histology were obtained. Our results indicate that topotecan shows non linear pharmacokinetics after a single intravitreal dose in the range of 0.5-5 μg in the rabbit. Vitreous concentration of lactone topotecan was close to the concentration assumed to be therapeutically active after 5 h of 0.5 μg intravitreal administration. Eyes injected with four weekly doses of topotecan (0.5 or 5 μg/dose) showed no significant differences in their ERG wave amplitudes and implicit times in comparison with control (p > 0.05). Animals showed no weight, hair loss or significant changes in hematologic values during the study period. There were no significant histologic damage of the retinas exposed to topotecan treatments. After intravitreal administration no topotecan could be detected in plasma during the follow-up period nor in the vitreous of treated and control animals after 1 week of the last injection. The present data shows that four weekly intravitreal injection of 5 μg of topotecan is safe for the rabbit eye. Despite multiple injections of 0.5 μg of topotecan are also safe to the rabbit eye, lactone topotecan vitreous concentrations were potentially active only after 5 h of the administration. We postulate promising translation to clinics for retinoblastoma treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T2705
Topotecan hydrochloride hydrate, ≥98% (HPLC and enzymatic)
C23H23N3O5 · xHCl · yH2O