EMAIL THIS PAGE TO A FRIEND
Clinical journal of the American Society of Nephrology : CJASN
Low plasma α-tocopherol concentrations and adverse clinical outcomes in diabetic hemodialysis patients.
PMID 23335039
Abstract
Trials with the antioxidant vitamin E have failed to show benefit in the general population. Considering the different causes of death in ESRD, this study investigated the association between plasma concentrations of α-tocopherol and specific clinical outcomes in diabetic hemodialysis patients. In 1046 diabetic hemodialysis patients (participants of the German Diabetes and Dialysis Study), α-tocopherol was measured in plasma by reversed-phase HPLC. By Cox regression analyses, hazard ratios were determined for prespecified end points according to baseline plasma α-tocopherol levels: sudden death (n=134), myocardial infarction (n=172), stroke (n=89), combined cardiovascular events (n=398), fatal infection (n=107), and all-cause mortality (n=508). Patients had a mean age of 66±8 years, and mean plasma α-tocopherol level was 22.8±9.6 µmol/L. Levels of α-tocopherol were highly correlated to triglycerides (r=0.63, P<0.001). Patients in the lowest α-tocopherol quartile had (in unadjusted analyses) a 79% higher risk of stroke and a 31% higher risk of all-cause mortality compared with patients in the highest quartile. The associations were attenuated after adjustment for confounders (hazard ratiostroke=1.56, 95% confidence interval=0.75-3.25; hazard ratiomortality=1.22, 95% confidence interval=0.89-1.69, respectively). There was no association between α-tocopherol and myocardial infarction, sudden death, or infectious death. Plasma α-tocopherol concentrations were not independently associated with cardiovascular outcomes, infectious deaths, or all-cause mortality in diabetic hemodialysis patients. The lack of association can partly be explained by a confounding influence of malnutrition, which should be considered in the planning of trials to reduce cardiovascular risk in dialysis patients.