EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Preparation of highly functionalised benzofurans from ortho-hydroxyphenones and dichloroethylene: applications and mechanistic investigations.
PMID 23335128
Abstract
Highly functionalised benzofurans have been prepared from ortho-hydroxyphenones and 1,1-dichloroethylene. The key intermediate, a chloromethylene furan, smoothly rearranged into the corresponding benzofuran carbaldehyde under acidic conditions. Some mechanistic investigations have been performed and several biologically active benzofurans have been synthesised.