EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Palladium-catalyzed asymmetric allylic alkylation of 3-aryloxindoles with allylidene dipivalate: a useful enol pivalate product.
PMID 23335176