EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Very bright europium complexes that stain cellular mitochondria.
PMID 23336102
Abstract
The synthesis, structure and photophysical properties of a series of highly emissive europium complexes is reported. Certain complexes enter mammalian cells by macropinocytosis and stain the mitochondria selectively, allowing observation of the Eu emission in cellulo by time-gated spectral imaging.