EMAIL THIS PAGE TO A FRIEND
Tidsskrift for den Norske laegeforening : tidsskrift for praktisk medicin, ny raekke
[Is the warfarin era over?].
PMID 23338065
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F9302
Factor X Activated (Xa) from bovine plasma, aqueous glycerol solution