EMAIL THIS PAGE TO A FRIEND
JAMA internal medicine
New studies about everyday types of chemical exposures: what readers should consider: comment on "A crossover study of noodle soup consumption in melamine bowls and total melamine excretion in urine".
PMID 23338122