EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
High Stokes shift perylene dyes for luminescent solar concentrators.
PMID 23338660
Abstract
Highly efficient plastic based single layer Luminescent Solar Concentrators (LSCs) require the design of luminophores having complete spectral separation between absorption and emission spectra (large Stokes shift). We describe the design, synthesis and characterization of a new perylene dye possessing Stokes shift as high as 300 meV, fluorescent quantum yield in the LSC slab of 70% and high chemical and photochemical stability.