EMAIL THIS PAGE TO A FRIEND
Naunyn-Schmiedeberg's archives of pharmacology
NDP-MSH inhibits neutrophil migration through nicotinic and adrenergic receptors in experimental peritonitis.
PMID 23338711
Abstract
Melanocortin is a potent anti-inflammatory molecule. However, little is known about the effect of melanocortin on acute inflammatory processes such as neutrophil migration. In the present study, we investigated the ability of [Nle4, D-Phe7]-melanocyte-stimulating hormone (NDP-MSH), a semisynthetic melanocortin compound, in the inhibition of neutrophil migration in carrageenin-induced peritonitis model. Herein, subcutaneous pretreatment with NDP-MSH decreased neutrophil trafficking in the peritoneal cavity in a dose-dependent manner. NDP-MSH inhibited vascular leakage, leukocyte rolling, and adhesion and reduced peritoneal macrophage inflammatory protein 2, but not TNF-alpha, IL-1beta, IL-10, and keratinocyte-derived chemokine production. In addition, the effect on neutrophil migration was reverted by the pretreatment with both propranolol (a nonselective beta-adrenergic antagonist) and mecamylamine (a nonselective nicotinic antagonist) but not by splenectomy surgery. Moreover, NDP-MSH intracerebroventricular administration inhibited neutrophil migration, indicating participation of the central nervous system. Our results propose that the NDP-MSH effect may be due to a spleen-independent neuro-immune pathway that efficiently regulates excessive neutrophil recruitment to tissues.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M8764
[Nle4, D-Phe7]-α-Melanocyte Stimulating Hormone trifluoroacetate salt, ≥95% (HPLC)
C78H111N21O19 · xC2HF3O2