EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Total synthesis of (-)-isoavenaciolide.
PMID 23339655
Abstract
An enantioselective approach to (-)-isoavenaciolide was achieved starting from 1-undecyn-3-ol. The synthesis relied upon the preparation of a chiral 4-silyloxy-2-alkenylborane by hydroboration of a protected 2,3-allenol and subsequent stereoselective addition to 2-thiophenecarboxaldehyde.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
20740
γ-Butyrolactone, puriss., ≥99.0% (GC)
C4H6O2