EMAIL THIS PAGE TO A FRIEND
Acta clinica Belgica
Taenia saginata a rare cause of acute cholangitis: a case report.
PMID 23340150
Abstract
Parasitic infestations of the galdbladder and biliary tract are quite rare. Taenia saginata is an intestinal helmint and patients harbouring adult T.saginata tapeworms are mostly asymptomatic and discharge only fecal proglottids. In some cases there might be nonspecific symptoms like vomiting, nausea, epigastric pain, diarrhea and weight loss. Tenia saginata is a also rare cause of ileus, pancreatitis, cholecystitis and cholangitis. We report a case of acute cholangitis caused by T. saginata presenting with fever, nausea, vomiting, jaundice and right upper quadrant pain. Although parasites are not an uncommon cause of cholangitis especially in diseaseendemic areas like the Far East, this is not true for T. saginata causing acute cholangitis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N3510
Niclosamide
C13H8Cl2N2O4
N0560000
Niclosamide (anhydrous), European Pharmacopoeia (EP) Reference Standard
C13H8Cl2N2O4