EMAIL THIS PAGE TO A FRIEND
The Analyst
Extraction, separation and spectrophotometric determination of cadmium and mercury using triphenylphosphine oxide and its application to environmental samples.
PMID 2334057
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T84603
Triphenylphosphine oxide, 98%
C18H15OP