EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Urea-functionalized crystalline capsules for recognition and separation of tetrahedral oxoanions.
PMID 23340607
Abstract
The persistent ability of tripodal TREN-based tris-urea receptors (TREN = tris(2-aminoethyl)amine) to self-assemble with a variety of oxoanions into dimeric capsules upon crystallization is reviewed. The capsule crystallization allows for charge-, shape-, and size-selective encapsulation of tetrahedral XO(4)(n-) anions (n = 2,3), and provides an effective way to separate these anions from competitive aqueous environments.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
225630
Tris(2-aminoethyl)amine, 96%
C6H18N4
93303
Tris(2-aminoethyl)amine, technical, ≥90% (GC)
C6H18N4