EMAIL THIS PAGE TO A FRIEND
Molecular diversity
An efficient and facile synthesis of 3-amino-5-chromenyl-butenolides from 3-formyl chromone, dialkyl acetylenedicarboxylate, and primary amines.
PMID 23341007
Abstract
A three-component reaction of 3-formyl chromones, dialkyl acetylenedicarboxylate, and isocyanides in the presence of POCl(3) was used as a highly efficient and practical approach for the synthesis of 3-amino-5-chromenyl-butenolides. High yields and high bond forming efficiency, and simple operations are the advantages of this method. Graphical abstract.