EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Supra-amphiphiles formed by complexation of azulene-based amphiphiles and pyrene in aqueous solution: from cylindrical micelles to disklike nanosheets.
PMID 23341054
Abstract
We have fabricated supra-amphiphiles by charge-transfer complexation of azulene-based amphiphiles and pyrene in water, and the self-assembled nanostructures can reversibly change between cylindrical micelles and disklike nanosheets in response to interaction with guest molecules.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
37879
Azulene, analytical standard
C10H8
A97203
Azulene, 99%
C10H8