EMAIL THIS PAGE TO A FRIEND
Chemistry & biodiversity
Synthesis and evaluation of gambogic acid derivatives as antitumor agents. Part III.
PMID 23341209
Abstract
Gambogic acid (GA) has been reported as a potent apoptosis inducer. Previously, we have reported chemical modification at C(34) and C(39) of GA, leading to some agents with improved activity. To investigate the further structure-activity relationship (SAR) and preliminary mechanism of GA activity, a series of derivatives with modified prenyl side chains of GA were synthesized and evaluated. Most of the derivatives showed potent inhibitory activities against the proliferation of HepG2 and A549 cell lines. Compound 4 was selected for further mechanistic studies due to its outstanding activity. It was established that 4 induces the apoptosis of HepG2 cells by using Annexin-V/PI double staining and Western blot assay, thus, compound 4 can serve as a promising lead compound for the development of novel apoptosis in anticancer treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G8171
Gambogic acid, ≥95% (HPLC), powder
C38H44O8