EMAIL THIS PAGE TO A FRIEND
Stem cells (Dayton, Ohio)
Efficient generation of astrocytes from human pluripotent stem cells in defined conditions.
PMID 23341249
Abstract
Astrocytes can be generated from various tissue sources including human pluripotent stem cells (PSC). In this manuscript, we describe a chemically defined xeno-free medium culture system for rapidly generating astrocytes from neural stem cells derived from PSC. We show that astrocyte development in vitro, mimics normal development in vivo, and also passes through a CD44(+) astrocyte precursor stage. Astrocytes generated by our method display similar gene expression patterns, morphological characteristics and functional properties to primary astrocytes, and they survive and integrate after xenotransplantation. Whole genome expression profiling of astrocyte differentiation was performed at several time points of differentiation, and the results indicate the importance of known regulators and identify potential novel regulators and stage-specific lineage markers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AST1001
Astrocytes, Progenitor, CTRL Line, IPSC derived