EMAIL THIS PAGE TO A FRIEND
Urologiia (Moscow, Russia : 1999)
[Application of alpha1-adrenoblockers in treatment of ureteral calculi].
PMID 23342609
Abstract
The results of evaluation of the efficacy of alpha1-adrenoblockers in treatment of ureteral calculi are presented. Comparative, prospective, placebo-uncontrolled nonrandomized single-center study was performed, which included 118 patients with solitary diagnosed calculi in various parts of ureter. After the pain relief, all the patients underwent conservative therapy aimed at spontaneous discharge of concretions. The maximum duration of conservative treatment was 28 days. Ultrasound monitoring was performed every week in all patients. The control group of patients received only Drotaverinum 40 mg three times a day, and analgesics. The main group received alpha1-adrenoblocker tamsulosin at a standard dose of 0.4 mg once a day along with Drotaverinum and analgesics. The overall probability of a discharge of concrements localized in the distal ureter was significantly (P = 0.02) higher in the patients treated with alpha1-adrenoblockers. Treatment regimen in main group of patients allowed better control of pain during all periods of observation, even if the discharge of concretions was not registered. The overall probability of migration of concrements from the proximal to the distal ureter in main group of patients was 52% versus 32% in controls (P = 0.17). The frequency of adverse effects was comparable in both groups. Vertigo, postural hypotension, and weakness were significantly more frequent in the main group of patients. Univariate and multivariate analyses of the proportional hazards model have demonstrated that the administration of alpha1-adrenoblocker increased the likelihood of a discharge of concrement from the distal ureter. It is shown that the nature of the applied therapy has directly influence on the risk of an earlier discharge of concretions. Inclusion of alpha1-adrenoblockers in the treatment scheme increased the probability of discharge of concrements at 4.11 times.