EMAIL THIS PAGE TO A FRIEND
Ukrains'kyi biokhimichnyi zhurnal (1999 )
[The influence of the penetrating cryoprotectors on protein synthesis in the cell-free system of the rat liver].
PMID 23342637
Abstract
The influence of the penetrating cryoprotector and Mg2+ ions on the protein-synthesizing activity of postmitochondrial supernantant of the rat liver as well as on aminoacylation processes has been investigated. The addition of the penetrating cryoprotectors--ethylene glycol and DMSO--resulted in the concentration-dependant reversible inhibition of the protein biosynthesis and aminoacylation reaction in the cell-free system. These cryoprotectors at low concentrations intensified the stimulating effect of Mg2+ on the cumulative protein synthesis in the cell-free system.
Related Materials