EMAIL THIS PAGE TO A FRIEND
The journal of physical chemistry. B
Unimolecular reactivity of the [urea-Sr]2+ complex, a metastable dication in the gas phase: an experimental and theoretical perspective.
PMID 23343443
Abstract
The interactions between urea and Sr(2+) in the gas phase have been investigated by combining electrospray ionization/mass spectrometry techniques and density functional and high-level ab initio molecular orbital calculations. Our theoretical survey indicates that [Sr(urea)](2+) adducts are thermodynamically stable with respect to direct Coulomb explosions. However, after isomerization, some of the local minima of the PES are thermodynamically unstable with respect to the formation of NH(4)(+), but kinetically metastable. The loss of neutral fragments with the concomitant generation of lighter doubly charged fragment ions, namely, [(H(3)N)Sr](2+) and [(HNCO)]Sr(2+), compete with the aforementioned Coulomb explosion processes yielding NH(4)(+) + [(NCO)Sr](+) and [(H(2)N)Sr](+) + [H(2)NCO](+), although the former processes dominate. Hence, both singly and doubly charged species are detected as dissociation products. Quite importantly, the observed eliminations of NH(3) or HNCO lead to the formation of new doubly charged species, which turn out to be thermodynamically stable.