EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Diastereoselective synthesis of 5-hydroxy-8-methoxy-1-oxaspiro[5,5]undeca-7,10-diene-9-one.
PMID 23344206
Abstract
A short five steps synthesis of the title compound from vanillin is described. The racemic spiroether 7 was obtained in 61% yield and in >99% diastereomeric excess (by 1H-NMR) from the corresponding phenolic derivative 3 by oxidation with lead (IV) acetate.