EMAIL THIS PAGE TO A FRIEND
ChemSusChem
Cyanomethylbenzoic acid: an acceptor for donor-π-acceptor chromophores used in dye-sensitized solar cells.
PMID 23345047
Abstract
Sensing the sun: Incorporation of a cyanomethyl benzoic acid electron acceptor into donor-π-acceptor sensitizers for dye-sensitized-solar cell is shown to lead to devices with improved conversion efficiency when compared with more widely used cyanoacetic acid acceptor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
290009
Potassium benzoate, ReagentPlus®, 99%
C7H5KO2