EMAIL THIS PAGE TO A FRIEND
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme
Androgen synthesis in patients with congenital adrenal hyperplasia due to 21-hydroxylase deficiency.
PMID 23345132
Abstract
A hallmark of severe congenital adrenal hyperplasia due to 21-hydroxylase deficiency is pre- and postnatal virilization. The most characteristic biochemical abnormality is the elevation of 17α-hydroxyprogesterone, which is metabolized to the most potent androgen receptor agonist dihydrotestosterone. 17α-Hydroxyprogesterone can be metabolized to dihydrotestosterone via 4-androstenedione through the classical Δ⁴-pathway or via 17α-hydroxypregnenolone and dehydroepiandrosterone through the classical Δ⁵-pathway, as well as through an alternative route, called the 'backdoor pathway', that bypasses dehydroepiandrosterone, 4-androstenedione, and testosterone as intermediates. This review article will summarize recent advances in the understanding of the activities of androgen synthesis pathways in patients with 21-hydroxylase deficiency obtained by urinary steroid metabolomics based on gas chromatography-mass spectrometry. Compared with healthy controls, the relative activities of the backdoor and Δ⁴-pathways increase in patients with congenital adrenal hyperplasia during neonatal age and infancy, whereas the activity of the Δ⁵-pathway remains unchanged. Thereafter, the activity of the Δ⁵-pathway dominates, whereas a decreasing 5α-reductase activity leads to a diminished role of the backdoor pathway for androgenic steroid production. Beside the backdoor pathway, the Δ⁴-pathway seems to be responsible for increased androgen generation in patients with 21-hydroxylase deficiency before the onset of adrenarche, whereas the Δ⁵-pathway might contribute to the increased androgen formation in those patients only after the onset of adrenarche.