EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Depletion of cellular polyamines, spermidine and spermine, causes a total arrest in translation and growth in mammalian cells.
PMID 23345430
Abstract
The polyamines, putrescine, spermidine, and spermine, are essential polycations, intimately involved in the regulation of cellular proliferation. Although polyamines exert dynamic effects on the conformation of nucleic acids and macromolecular synthesis in vitro, their specific functions in vivo are poorly understood. We investigated the cellular function of polyamines by overexpression of a key catabolic enzyme, spermidine/spermine N(1)-acetyltransferase 1 (SAT1) in mammalian cells. Transient cotransfection of HeLa cells with GFP and SAT1 vectors suppressed GFP protein expression without lowering its mRNA level, an indication that the block in GFP expression was not at transcription, but at translation. Fluorescence single-cell imaging also revealed specific inhibition of endogenous protein synthesis in the SAT1 overexpressing cells, without any inhibition of synthesis of DNA or RNA. Overexpression of SAT1 using a SAT1 adenovirus led to rapid depletion of cellular spermidine and spermine, total inhibition of protein synthesis, and growth arrest within 24 h. The SAT1 effect is most likely due to depletion of spermidine and spermine, because stable polyamine analogs that are not substrates for SAT1 restored GFP and endogenous protein synthesis. Loss of polysomes with increased 80S monosomes in the polyamine-depleted cells suggests a direct role for polyamines in translation initiation. Our data provide strong evidence for a primary function of polyamines, spermidine and spermine, in translation in mammalian cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
05292
Spermidine 0.1 M solution