EMAIL THIS PAGE TO A FRIEND
The European Journal of Neuroscience
Viral transduction of the neonatal brain delivers controllable genetic mosaicism for visualising and manipulating neuronal circuits in vivo.
PMID 23347239