EMAIL THIS PAGE TO A FRIEND
Eye (London, England)
Intraorbital corticosteroid injection for orbital reactive lymphoid hyperplasia.
PMID 23348726
Abstract
The purpose of this study is to assess the utility of intraorbital injection of triamcinolone acetonide as a primary treatment option for orbital reactive lymphoid hyperplasia (RLH). Retrospective, single-centre, interventional case series. Intraorbital injection of triamcinolone acetonide was associated with complete resolution of all symptoms and signs in four cases (80%). This was achieved with a single injection in two cases and with two injections in another two cases. Radiological resolution was confirmed in one case. One case developed bilateral multifocal orbital RLH lesions 1 month after the second injection. Intraorbital injection of corticosteroid may be a useful treatment option for orbital RLH, and may have a role as a first-line therapy in RLH of the anterior orbit. A significant proportion of patients may require repeat injections to achieve resolution. A larger prospective study is required to validate our findings.