EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Crystal structure, antibacterial and cytotoxic activities of a new complex of bismuth(III) with sulfapyridine.
PMID 23348999
Abstract
A new complex of Bi(III) and sulfapyridine was synthesized and characterized by elemental analysis, atomic absorption spectrometry, conductivity analysis, electrospray ionization mass spectrometry (ESI-MS), infrared spectroscopy, and single crystal X-ray diffraction methods. The antimicrobial and the cytotoxic activities of the compound were investigated. Elemental and conductivity analyses are in accordance to the formulation [BiCl₃(C₁₁H₁₁N₃O₂S)₃]. The structure of the complex reveals a distorted octahedral geometry around the bismuth atom, which is bound to three sulfonamidic nitrogens from sulfapyridine, acting as a monodentate ligand, and to three chloride ions. The presence of the compound in solution was confirmed by ESI-MS studies. The complex is 3 times more potent than the ligand against Salmonella typhimurium, 4 times against Staphylococcus aureus, Shigella dysenteriae, and Shigella sonnei and 8 times more potent against Pseudomonas aeruginosa and Escherichia coli. The compound inhibits the growth of chronic myelogenous leukemia cells with an IC₅₀ value of 44 μM whereas the free ligand has no effect up to 100 μM.