EMAIL THIS PAGE TO A FRIEND
Annals of the rheumatic diseases
Changing sulphasalazine to methotrexate does not improve the 2-year outcomes of the initial single DMARD treatment in early rheumatoid arthritis: subanalysis of the FIN-RACo trial.
PMID 23349132