PloS one
Growth and spectral assessment of Yb(3+)-doped KBaGd(MoO4)3 crystal: a candidate for ultrashort pulse and tunable lasers.
PMID 23349892
Abstract
In order to explore new more powerful ultrashort pulse laser and tunable laser for diode-pumping, this paper reports the growth and spectral assessment of Yb(3+)-doped KBaGd(MoO(4))(3) crystal. An Yb(3+):KBaGd(MoO(4))(3) crystal with dimensions of 50×40×9 mm(3) was grown by the TSSG method from the K(2)Mo(2)O(7) flux. The investigated spectral properties indicated that Yb(3+):KBaGd(MoO(4))(3) crystal exhibits broad absorption and emission bands, except the large emission and gain cross-sections. This feature of the broad absorption and emission bands is not only suitable for the diode pumping, but also for the production of ultrashort pulses and tunability. Therefore, Yb(3+):KBaGd(MoO(4))(3) crystal can be regarded as a candidate for the ultrashort pulse and tunable lasers.