EMAIL THIS PAGE TO A FRIEND
Journal de pharmacie de Belgique
[Racecadotril (Tiorfix), oral].
PMID 23350211
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T6031
DL-Thiorphan, ≥98% (TLC)
C12H15NO3S
SML0043
Racecadotril, ≥98% (HPLC)
C21H23NO4S