EMAIL THIS PAGE TO A FRIEND
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.]
Polycaprolactone for the correction of nasolabial folds: a 24-month, prospective, randomized, controlled clinical trial.
PMID 23350617
Abstract
In this study, we examined two polycaprolactone (PCL)-based dermal filler formulas (PCL-1; PCL-2) for safety, patient satisfaction, likelihood to return, efficacy, and duration of correction. This 40-patient, 24-month, prospective, randomized, controlled study evaluated the efficacy, safety, longevity, and volume of two PCL formulas for correction of nasolabial folds. Patients enrolled in a medical clinic in Europe received two injections 1 month apart and returned at 3, 6, 9, 12, 15, 18, and 24 months for blinded patient evaluation using accepted aesthetic rating scales. At 12 months, the efficacy outcomes on Wrinkle Severity Rating Scale (WSRS) and Global Aesthetic Improvement Scale (GAIS) of PCL-1 and PCL-2 were consistently maintained, with sustained improvement in 90% and 91.4% of patients, respectively. At 24 months, PCL-2 was found to be more effective than PCL-1 with respect to GAIS and WSRS, showing sustained improvement for the entire 2-year study period (linear p = .52; quadratic p > .99). Patient satisfaction at 24 months was 72.4% for PCL-1 and 81.7% for PCL-2. Both products were found to be safe and well tolerated. PCL-1 and PCL-2 are safe and have sustained efficacy and high patient satisfaction, with PCL-2 demonstrating longer-lasting results than PCL-1.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
704105
Polycaprolactone, average Mn 45,000