EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Polyamino geranic derivatives as new chemosensitizers to combat antibiotic resistant gram-negative bacteria.
PMID 23352753
Abstract
Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate reagent (BOP) serves as an efficient and versatile coupling reagent for the design and synthesis of new polyamino geranic acid derivatives in moderate to good chemical yields varying from 47% to 83%. These compounds induced a significant decrease of antibiotic resistance in two Gram-negative bacterial MDR strains. Our data suggested that their mechanism of action is closely associated with the inhibition of the efflux pumps.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
427764
Geranic acid, technical grade, 85%
C10H16O2
W412101
Geranic acid, 85%, stabilised
C10H16O2