EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Synthesis, experimental, theoretical characterization and biological activities of 4-ethyl-5-(2-hydroxyphenyl)-2H-1,2,4-triazole-3(4H)-thione.
PMID 23352922
Abstract
This work presents the characterization of 4-ethyl-5-(2-hydroxyphenyl)-2H-1,2,4-triazole-3(4H)-thione (III) by quantum chemical calculations and spectral techniques. The molecular geometry, vibrational frequencies and gauge including atomic orbital (GIAO) (1)H and (13)C NMR chemical shift values of III in the ground state have been calculated using the density functional method (B3LYP) with the 6-31G(d) basis set. The calculated results show that the optimized geometry can well reproduce the crystal structure, and the theoretical vibrational frequencies and chemical shift values show good agreement with experimental values. To determine conformational flexibility, the molecular energy profile of the title compound was obtained by DFT calculations with respect to the selected torsion angle, which was varied from -180° to +180° in steps of 10°. The energetic behavior of III in solvent media was examined using the B3LYP method with the 6-31G(d) basis set by applying the Onsager and the polarizable continuum model (PCM). The predicted nonlinear optical properties of III are greater than ones of urea. In addition, DFT calculations of molecular electrostatic potentials and frontier molecular orbitals of III were carried out at the B3LYP/6-31G(d) level of theory. The title compound was screened for antibacterial, antifungal and antioxidant activities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T46108
1,2,4-Triazole, 98%
C2H3N3