EMAIL THIS PAGE TO A FRIEND
Life sciences
Inhibition by menthol and its related chemicals of compound action potentials in frog sciatic nerves.
PMID 23352972
Abstract
Transient receptor potential (TRP) vanilloid-1 (TRPV1) and melastatin-8 (TRPM8) channels play a role in transmitting sensory information in primary-afferent neurons. TRPV1 agonists at high concentrations inhibit action potential conduction in the neurons and thus have a local anesthetic effect. The purpose of the present study was to know whether TRPM8 agonist menthol at high concentrations has a similar action and if so whether there is a structure-activity relationship among menthol-related chemicals. Compound action potentials (CAPs) were recorded from the frog sciatic nerve by using the air-gap method. (-)-Menthol and (+)-menthol concentration-dependently reduced CAP peak amplitude with the IC(50) values of 1.1 and 0.93 mM, respectively. This (-)-menthol activity was resistant to non-selective TRP antagonist ruthenium red; TRPM8 agonist icilin did not affect CAPs, indicating no involvements of TRPM8 channels. p-Menthane, (+)-limonene and menthyl chloride at 7-10 mM minimally affected CAPs. On the other hand, (-)-menthone, (+)-menthone, (-)-carvone, (+)-carvone and (-)-carveol (in each of which chemicals OH or O group was added to p-menthane and limonene) and (+)-pulegone inhibited CAPs with extents similar to that of menthol. 1,8-Cineole and 1,4-cineole were less effective while thymol and carvacrol were more effective than menthol in inhibiting CAPs. Menthol-related chemicals inhibited CAPs and were thus suggested to exhibit local anesthetic effects comparable to those of lidocaine and cocaine as reported previously for frog CAPs. This result may provide information to develop local anesthetics on the basis of the chemical structure of menthol.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
63670
(±)-Menthol, racemic, ≥98.0% (GC)
C10H20O
82569
(+)-Pulegone, analytical standard
C10H16O
192384
(−)-Carveol, mixture of isomers, 97%
C10H16O
61370
(−)-Carveol, mixture of isomers, analytical standard
C10H16O
62118
(R)-(+)-Limonene, analytical standard
C10H16
62120
(R)-(+)-Limonene, purum, ≥96.0% (sum of enantiomers, GC)
C10H16
62122
(R)-(+)-Limonene, technical, ~90% (sum of enantiomers, GC)
C10H16
183164
(R)-(+)-Limonene, 97%
C10H16
376388
(R)-(+)-Pulegone, 97%
C10H16O
P55708
(R)-(+)-Pulegone, 85%, technical grade
C10H16O
W296309
(R)-(+)-Pulegone, ≥90%
C10H16O
62128
(S)-(−)-Limonene, analytical standard
C10H16
W504505
(S)-(−)-Limonene, ≥95%, FG
C10H16
218367
(S)-(−)-Limonene, 96%
C10H16
328847
(S)-(−)-Pulegone, 98%
C10H16O
27393
1,4-Cineole, analytical standard
C10H18O
27395
1,4-Cineole, technical, mixture of isomers, ≥85% (GC)
C10H18O
00020590
1,8-Cineole, primary reference standard
C10H18O
W224502
Carvacrol, ≥98%, FCC, FG
C10H14O
282197
Carvacrol, 98%
C10H14O
42632
Carvacrol, analytical standard
C10H14O
W224511
Carvacrol, natural, 99%, FG
C10H14O
Y0000176
Cineole, European Pharmacopoeia (EP) Reference Standard
C10H18O
W266507
DL-Menthol, ≥95%, FCC, FG
C10H20O
29210
Eucalyptol, analytical standard
C10H18O
95656
Eucalyptol, certified reference material, TraceCERT®
C10H18O
C80601
Eucalyptol, 99%
C10H18O
W246506
Eucalyptol, natural, ≥99%, FCC, FG
C10H18O
W224707
L-Carveol, mixture of cis and trans, ≥95%, FG
C10H16O
M2772
Menthol, 99%
C10H20O