EMAIL THIS PAGE TO A FRIEND
International journal of oncology
FKBP10/FKBP65 expression in high-grade ovarian serous carcinoma and its association with patient outcome.
PMID 23354471
Abstract
The frequent loss of chromosome 17 in epithelial ovarian carcinomas (EOC), particularly high-grade serous carcinomas (HGSC), has been attributed to the disruption of TP53 (at 17p13.1) and other chromosome 17 genes suspected to play a role in tumour suppressor pathways. In a transcriptome analysis of HGSC, we showed underexpression of a number of chromosome 17 genes, which included FKBP10 (at 17q21.1) and collagen I α 1 (COL1A1; at 17q21.33). FKBP10 codes for the immunophilin FKBP65 and is suspected to act as a chaperone for COL1A1. We have investigated FKBP10 (gene) and FKBP65 (protein) expression in HGSC samples and EOC cell lines that differ in their tumourigenic potential. COL1A1 expression was also investigated given the purported function of FKBP65. RT-PCR analysis verified underexpression of FKBP10 and COL1A1 in HGSCs (n=14) and six tumourigenic EOC cell lines, relative to normal ovarian surface epithelial cells and a non-tumourigenic EOC cell line. Immunohistochemistry analyses of 196 HGSC samples using tissue microarrays revealed variable staining intensities in the epithelial tumour component where only 7.8% and 1.0% of samples stained intensely for FKBP65 and COL1A1, respectively. Variable staining intensities were also observed for the stromal component where 23.6% and 24.1% stained intensely for FKBP65 and COL1A1, respectively. There was no significant correlation of staining intensity of either protein with disease stage. Staining of FKBP65 was clearly visible in normal epithelial cells of the ovarian surface and fallopian tube. There was a significant correlation between absence of FKBP65 staining in the epithelial cell component of the tumour and prolonged overall survival (p<0.001). Our results suggest that underexpression of FKBP65 protein is characteristic of HGSCs and that this expression profile may be linked to molecular pathways associated with an unfavourable outcome in cancer patients.