EMAIL THIS PAGE TO A FRIEND
Environmental science and pollution research international
Joint action and lethal levels of toluene, ethylbenzene, and xylene on midge (Chironomus plumosus) larvae.
PMID 23354551
Abstract
Aquatic ecosystems are vulnerable to the exposure with petrochemicals such as toluene, ethylbenzene, and xylene (o-, m-, and p-xylene) (TEX) and their adverse effects. Considering the widespread use, occurrence, and high toxicity of TEX, the aim of this work was to investigate the differential toxicity of TEX against midge (Chironomus plumosus) larvae and reveal the joint action of binary and ternary mixtures of TEX using the predictive concentration addition model. More importantly, this research can afford the basic toxicity data and scientific reference for the establishment of water quality criteria or benchmark, water pollution control, and aquatic risk assessment. Single and joint toxic effects of TEX on C. plumosus larvae were investigated using a semi-static bioassay, and the type of joint effects of TEX was ascertained. In the single toxicant experiments, the toxicity of the three pollutants could be sequenced as ethylbenzene > xylene > toluene. Specifically, LC(50s) of T, E, and X after a 48-h exposure were 64.9, 37.8, and 42.0 mg/L, respectively. In the binary mixture experiments, the interaction between toluene and ethylbenzene, ethylbenzene and xylene, and toluene and xylene was largely in conformity with partial additive or additive effect as determined by isobologram representation and toxic unit models. In the ternary mixture experiments, the interaction was basically dependent on the use of additive index and mixture toxicity index methods. However, the antagonistic and synergistic actions were not significant. Thus, the tertiary mixture interaction could be regarded as additive action. The concentration addition model could successfully predict the joint action of TEX mixtures on C. plumosus larvae. Particularly, the additive action of TEX on C. plumosus larvae can be further recommended to evaluate water quality criteria of TEX.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
03079
Ethylbenzene, analytical standard
C8H10
E12508
Ethylbenzene, ReagentPlus®, 99%
C8H10
40036
Ethylbenzene solution, certified reference material, 5000 μg/mL in methanol
C8H10
612936
Ethylbenzene solution, NMR reference standard, 5% in chloroform-d (99.8 atom % D), TMS 2 %, NMR tube size 3 mm × 8 in.
C8H10
551341
Ethylbenzene solution, NMR reference standard, 5% in chloroform-d (99.8 atom % D), TMS 2 %, NMR tube size 5 mm × 8 in.
C8H10
487112
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 3 mm × 8 in.
C8H10
487104
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in.
C8H10
487120
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 8 mm × 8 in.
C8H10
612065
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 10 mm × 8 in.
C8H10
612480
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, chromium(III) acetylacetonate 0.1 mg/mL, NMR tube size 5 mm × 8 in.
C8H10
708046
Ethylbenzene solution, NMR reference standard, 10% in chloroform-d (99.8 atom % D)
C8H10
708054
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d ("100%", 99.96 atom % D)
C8H10
710725
Ethylbenzene solution, NMR reference standard, 5% in chloroform-d (99.8 atom % D), TMS 1 %, NMR tube size 5 mm × 7 in.
C8H10
714917
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in. , ultra-thin wall
C8H10
717967
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.1 %
C8H10
717959
Ethylbenzene solution, NMR reference standard, 1% in chloroform-d (99.8 atom % D)
C8H10
736198
Ethylbenzene solution, NMR reference standard, 10% in chloroform-d (99.8 atom % D), NMR tube size 5 mm × 8 in. , ultra-thin wall
C8H10
733768
Ethylbenzene solution, NMR reference standard, 10% in chloroform-d (99.8 atom % D), NMR tube size 10 mm × 8 in.
C8H10
721018
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), NMR tube size 5 mm × 8 in.
C8H10
721026
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), NMR tube size 3 mm × 8 in.
C8H10
720291
Ethylbenzene solution, NMR reference standard, 10% in chloroform-d (99.8 atom % D), NMR tube size 3 mm × 8 in.
C8H10
40084 Toluene solution, certified reference material, 5000 μg/mL in methanol