EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
Partial enzymatic elimination and quantification of sarcosine from alanine using liquid chromatography-tandem mass spectrometry.
PMID 23354581
Abstract
Since sarcosine and D,L-alanine co-elute on reversed-phase high-performance liquid chromatography (HPLC) columns and the tandem mass spectrometer cannot differentiate them due to equivalent parent and fragment ions, derivatization is often required for analysis of sarcosine in LC/MS systems. This study offers an alternative to derivatization by employing partial elimination of sarcosine by enzymatic oxidation. The decrease in apparent concentration from the traditionally merged sarcosine-alanine peak associated with the enzymatic elimination has been shown to be proportional to the total sarcosine present (R(2) = 0.9999), allowing for determinations of urinary sarcosine. Sarcosine oxidase was shown to eliminate only sarcosine in the presence of D,L-alanine, and was consequently used as the selective enzyme. This newly developed technique has a method detection limit of 1 μg/L (parts per billion) with a linear range of 3 ppb-1 mg/L (parts per million) in urine matrices. The method was further validated through spiked recoveries of real urine samples, as well as the analysis of 35 real urine samples. The average recoveries for low, middle, and high sarcosine concentration spikes were 111.7, 90.8, and 90.1 %, respectively. In conclusion, this simple enzymatic approach coupled with HPLC/MS/MS is able to resolve sarcosine from D,L-alanine leading to underivatized quantification of sarcosine.
Related Materials