EMAIL THIS PAGE TO A FRIEND
European journal of clinical pharmacology
Population pharmacokinetics of piperacillin/tazobactam in neonates and young infants.
PMID 23354809
Abstract
To develop population pharmacokinetic (PK) models for piperacillin/tazobactam in neonates and infants of less than 2 months of age in order to determine the appropriate dosing regimen and provide a rational basis for the development of preliminary dosing guidelines suitable for this population. A two-stage, open-label study was conducted in neonates and infants less than 2 months of age in the neonatal intensive care unit (NICU). A total of 207 piperacillin and 204 tazobactam concentration-time data sets from 71 patients were analyzed using a nonlinear mixed-effect modeling approach (NONMEM VII). PK models were developed for piperacillin and tazobactam. The final models were evaluated using both bootstrap and visual predictive checks. External model evaluations were made in 20 additional patients. For neonates and young infants less than 2 months of age, the median central clearance was 0.133 and 0.149 L/h/kg for piperacillin and tazobactam, respectively. Postmenstrual age (PMA) was identified as the most significant covariate on central clearance of piperacillin and tazobactam. However, the combination of current bodyweight (BW) and postnatal age proved to be superior to PMA alone. BW was the most important covariate for apparent central volume of distribution. Both internal and external evaluations supported the prediction of the final piperacillin and tazobactam PK models. The dosing strategy 44.44/5.56 mg/kg/dose piperacillin/tazobactam every 8 or 12 h evaluated in this study achieved the pharmacodynamic target (free piperacillin concentrations >4 mg/L for more than 50 % of the dosing interval) in about 67 % of infants. Population PK models accurately described the PK profiles of piperacillin/tazobactam in infants less than 2 months of age. The results indicated that higher doses or more frequent dosing regimens may be required for controlling infection in this population in NICU.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P8396
Piperacillin sodium salt, penicillin analog
C23H26N5NaO7S