EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Triggering cell adhesion, migration or shape change with a dynamic surface coating.
PMID 23355329
Abstract
There's an APP for that: cell-repellent APP (azido-[polylysine-g-PEG]) is used to create substrates for spatially controlled dynamic cell adhesion. The simple addition of a functional peptide to the culture medium rapidly triggers cell adhesion. This highly accessible yet powerful technique allows diverse applications, demonstrated through tissue motility assays, patterned coculturing and triggered cell shape change.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8052
Arg-Gly-Asp, ≥97% (TLC)
C12H22N6O6