EMAIL THIS PAGE TO A FRIEND
Otolaryngology--head and neck surgery : official journal of American Academy of Otolaryngology-Head and Neck Surgery
Despite warnings, some graduating otolaryngology residents planning to use codeine for posttonsillectomy pain control.
PMID 23355375